Learn More
Invitrogen™ Substance P Competitive ELISA Kit
Click to view available options
Cantidad:
96 pruebas
Tamaño de la unidad:
paquete de 96
Descripción
Substance P (SP) is currently thought to be one of the most important neurotransmitters and neurotransmitters in the brain. It is an undecapeptide belonging to the tachykinin family, which consists of excitatory neuropeptides synthesised in neuronal and glial cells of the human central and peripheral nervous system. Substance P, Neurokinin A, Neuropeptide K, and Neuropeptide gamma are all generated by post-translation cleavage of the precursor Protachykinin-1. Substance P forms the major endogenous ligand for Neurokinin 1 Receptor. The pharmacology of Substance P has been associated with a number of neurological and psychiatric disorders, namely nociception, migraine, asthma, nausea, inflammatory bowel syndrome, urinary incontinence, anxiety and depression. It has also been linked to obesity.
Especificaciones
Especificaciones
| Intervalo del ensayo | De 78,13 a 5000 pg/ml |
| Sensibilidad del ensayo | 46.88 pg/mL |
| Conjugado | HRP |
| Tipo de producto | ELISA |
| Tipo de muestra | Plasma, Serum |
| Para utilizar con (equipo) | Lector de microplacas colorimétrico |
| CV interensayo | <10% |
| CV intraensayo | <10 |
| Contenido del kit | Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer |
| Etiqueta o tinte | HRP |
| Mostrar más |