missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STRO-1 Monoclonal Antibody (STRO-1), PE

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA528636
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody recognizes an extracellular epitope of the cell surface antigen STRO-1 expressed by bone marrow mesenchymal stromal cells and nucleated erythroid precursors, but not by committed hematopoietic progenitors.
STRO-1 is a cell surface antigen expressed by stromal elements in human bone marrow. STRO-1 binds to approximately 10% of bone marrow mononuclear cells, greater than 95% of which are nucleated erythroid precursors, but does not react with committed progenitor cells (colony-forming unit granulocyte-macrophage (CFU-GM), erythroid bursts (BFU-E), and mixed colonies (CFU-Mix). From bone marrow cells, the frequency of fibroblast colony forming cells (CFU-F) is enriched approximately 100-fold in the STRO-1-positive/Glycophorin A-negative population than in the STRO-1-positive/Glycophorin A-positive population. When plated under long-term BM culture conditions, STRO-1-positive cells generate adherent cell layers containing multiple stromal cell types, including adipocytes, smooth muscle cells, and fibroblastic elements. A STRO-1-positive enriched subset of marrow cells is capable of differentiating into multiple mesenchymal lineages including hematopoiesis-supportive stromal cells with a vascular smooth muscle-like phenotype, adipocytes, osteoblasts and chondrocytes. STRO-1 is a valuable antibody for the identification, isolation and functional characterization of human bone marrow stromal cell precursors.
Especificaciones
| STRO-1 | |
| Monoclonal | |
| 0.1 mg/mL | |
| TBS with 0.2% BSA and 15mM sodium azide; pH 8 | |
| Mouse | |
| Size-exclusion chromatography | |
| RUO | |
| Human, Rat, Rabbit | |
| Antibody | |
| IgM λ |
| Flow Cytometry | |
| STRO-1 | |
| PE | |
| STRO1 | |
| Human CD34 positive bone marrow cells. | |
| 100 μg | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido