missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Streptococcus Group A Monoclonal Antibody (HSA12-322.2), Invitrogen™
Descripción
MA1-10699 detects Streptococci Group A. MA1-10699 has been successfully used in ELISA applications. The MA1-10699 immunogen is UV-inactivated Group A Streptococcus cells.
The genus Streptococcus is comprised of a variety of pathogenic and commensal bacteria with broad importance in both medicine and industry. The genus Streptococcus is comprised of a wide variety of both pathogenic and commensal gram positive bacteria which are found to inhabit a wide range of hosts, including humans, horses, pigs and cows. Within the host, streptococci are often found to colonize the mucosal surfaces of the mouth, nares and pharynx. However, in certain circumstances, they may also inhabit the skin, heart or muscle tissue. Streptococci are often important as part of animal natural flora, but under the right circumstance can cause disease. Streptococci are classified largely according to the various serogroups. Group A and B streptococci have particular importance in medicine as they can cause severe infections.
Especificaciones
Especificaciones
| Antígeno | Streptococcus Group A |
| Aplicaciones | ELISA |
| Clasificación | Monoclonal |
| Clon | HSA12-322.2 |
| Concentración | 0.9 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative; pH 7.4 |
| Especie del huésped | Mouse |
| Inmunógeno | UV-inactivated Group A Streptococcus cells. |
| Método de purificación | Protein G |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?