Learn More
Riley™ Stream Evo Eco LED Safety Glasses
Descripción
- Stream Evo Eco Safety Glasses Provide Extreme Comfort and Wearability with a Range of Lens Options to Suit a Variety of Conditions
- Single wraparound polycarbonate lens
- Soft
- Available in a smaller size - Stream Evo Small
- No single use packaging
Especificaciones
Especificaciones
| Revestimiento de lente | Scratch-resistant, Anti-fog |
| Tipo de producto | Safety Glasses |
| Color de marco | Gray |
| N.º por caja | 200/Cs. |
| Peso (imperial) | 2.38 oz. |
| Peso (métrico) | 67.5 g |
| Material | Polycarbonate |
| Desechables | Reusable |
| Lens Material | Polycarbonate |
| Material del bastidor | Polycarbonate |
| Mostrar más |