missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STK11IP Polyclonal Antibody

Código de producto. 11356973
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11356973 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11356973 Supplier Invitrogen™ Supplier No. PA518996

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.

May regulate STK11/LKB1 function by controlling its subcellular localization.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno STK11IP
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
génica STK11IP
N.º de referencia del gen Q8N1F8
Alias de gen 1200014D22Rik; BB131189; KIAA1898; LIP1; LKB1-interacting protein 1; LKB1IP; serine/threonine kinase 11 interacting protein; serine/threonine kinase 11-interacting protein; serine/threonine-protein kinase 11-interacting protein; STK11 interacting protein; STK11IP; STK11IP1
Símbolos de los genes STK11IP
Especie del huésped Goat
Inmunógeno Synthetic peptide sequence (SRGTPNRERKQGEQS) corresponding to the internal amino acids of STK11IP (aa 721-736).
Método de purificación Ammonium sulfate precipitation
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 114790
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.