missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STING/TMEM173 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-24683AF594
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
STING/TMEM173 Polyclonal specifically detects STING/TMEM173 in Human, Mouse, Canine, Primate, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
| STING/TMEM173 | |
| Polyclonal | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, Stimulator of interferon genes protein, sting, STINGhSTING, TMEM173, transmembrane protein 173 | |
| Rabbit | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Canine, Primate, Rhesus Macaque | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TMEM173 | |
| Partial synthetic peptide made to an internal portion of human STING/TMEM173 (between amino acids 310-360) [UniProt Q86WV6] | |
| Cancer, Immunology, Innate Immunity | |
| 340061 | |
| Store at 4°C in the dark. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido