Learn More
KARTELL Sterile Petri Dish
Descripción
- Disposable planar, clear Petri dishes in PS.
- Ventilated and radiation- sterilized.
- Designed for routine lab work, bacteriological research.
- CE Marked.
- With aeration notches on the lid to prevent condensation formation.
- Beta-ray sterilization
- Designed for routine lab work, bacteriological research.
- Gas exchange outside Petri dish is reduced thanks to the absence of notches.
Especificaciones
Especificaciones
| Esterilizables en autoclave | No autoclavable |
| Forma del fondo | Plano |
| Color | Clear |
| Desechables | Single-use |
| Cuadriculado | Not Gridded |
| Material | Poliestireno |
| Forma | Circular |
| Esterilidad | Estéril |
| Tratamiento de superficie | Sin tratar |
| Tipo | Petri Dish |
| Mostrar más |