missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT3, p Ser727 Antibody (788335), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals FAB4934H
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
STAT3 Monoclonal antibody specifically detects STAT3 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining)
Especificaciones
| STAT3 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining) | |
| 788335 | |
| Western Blot, Intracellular Staining by Flow Cytometry | |
| Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor) | |
| Phosphopeptide containing the human STAT3 S727 site | |
| 0.1 mL | |
| Cellular Markers, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6774 | |
| Store at 4°C in the dark. | |
| IgG2b |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido