Learn More
Invitrogen™ Staphylococcus aureus PBP2a (Methicillin-resistant) Monoclonal Antibody (AC10), Biotin
GREENER_CHOICE
Mouse Monoclonal Antibody
Marca:  Invitrogen™ MA546569
Código de producto. 17820226
  • 379.00€ / 50 microgramos
Descripción
Staphylococcus aureus PBP2a (Methicillin-resistant) Monoclonal Antibody for ICC/IF, ELISA
Methicillin-resistant Staphylococcus aureus (MRSA) is a type of bacteria that is resistant to the antibiotic methicillin. The resistance is due to the presence of a protein called penicillin-binding protein 2a (PBP2a), which is encoded by the mecA gene. PBP2a has a low affinity for methicillin and other beta-lactam antibiotics, which allows the bacteria to survive and multiply in the presence of these drugs. A target description for PBP2a would be a transpeptidase enzyme that is responsible for the synthesis of the bacterial cell wall and is a key factor in the resistance of MRSA to beta-lactam antibiotics.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Staphylococcus aureus PBP2a (Methicillin-resistant)
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
Mouse
Protein A
RUO
Bacteria
Antibody
IgG2a
ELISA, Immunocytochemistry
AC10
Biotin
MRSA
Native Staphylococcus aureus (methicillin resistant).
50 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones