Learn More
Gilac™ Stainless Steel Trolley for Large Volume Container
Descripción
- Trolley equiped with 125 mm diameter wheels in white PA (2 fixed and 2 pivoting with brakes).
- Stainless steel recommended in humid environments (no damage to bearings even in the presence of water).
- Makes it easier to work at breast height.
Especificaciones
Especificaciones
| Tipo | Carro |
| Admite | Large Volume Containers |
| Capacidad portante (métrico) | 150 kg |
| Material | Acero inoxidable |
| Diámetro de las ruedas (métrico) | 125 mm |
| Para utilizar con (equipo) | Recipiente semicircular de 220 L |
| Altura (imperial) | 35.4 in. |
| Altura (métrico) | 900 mm |