Learn More
Bochem™ Tapa de acero inoxidable
Código de producto. 10005250
Click to view available options
Para utilizar con (equipo):
Para bidón de medición graduado de 10 l
Para cubo graduado de 10 l
Para cubo graduado de 15 l
Para jarra graduada de 15 l
Unit Size:
cada uno
This item is not returnable. View return policy
Product Code. 10005250
Brand: Bochem™ 8332
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.