Descripción
- Telescopic samplers are often used for taking surface samples from rivers, tanks and lakes
- A wide choice of attachments is available to make the TeleScoop ideally suited for most applications
- Attachments are interchangeable and simply click into position
Especificaciones
Especificaciones
| Tipo de producto | Cacillo |
| Material | Acero inoxidable |
| Esterilizables en autoclave | Autoclavable |
| Capacidad (métrico) | 500 mL |
| Desechables | Reusable |
| Graduado | No |
| Esterilidad | No estéril |