missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ST3GAL1 Polyclonal Antibody

Descripción
Recommended positive controls: HeLa. Predicted reactivity: Rhesus Monkey (97%), Chimpanzee (99%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ST3GAL1 is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. It is normally found in the Golgi but can be proteolytically processed to a soluble form. Correct glycosylation of ST3GAL1 protein may be critical to its sialyltransferase activity. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4B.The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein is normally found in the Golgi but can be proteolytically processed to a soluble form. Correct glycosylation of the encoded protein may be critical to its sialyltransferase activity. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4B. Two transcript variants encoding the same protein have been found for this gene. Other transcript variants may exist, but have not been fully characterized yet.
Especificaciones
Especificaciones
| Antígeno | ST3GAL1 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with no preservative; pH 7 |
| génica | ST3GAL1 |
| N.º de referencia del gen | Q11201 |
| Alias de gen | 5330418N22Rik; AI467004; Alpha 2,3-ST 1; alpha-2,3-sialyltransferase ST3Gal I-r1; alpha-2-3-sialyltransferase; beta-galactoside alpha-2,3-sialyltransferase 1; CMP-N-acetylneuraminate: [beta-galactosidase alpha-2,3] sialytransferase; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 1; Gal beta1,3 GalNAc alpha2,3 sialyltransferase; Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase; Gal-NAc6S; MGC9183; putative alpha-2,3-sialyltransferase; Sialyltransferase 4A; sialyltransferase 4A (beta-galactosidase alpha-2,3-sialytransferase); sialyltransferase 4A (beta-galactoside alpha-2,3-sialyltransferase); sialyltransferase 4A (beta-galactoside alpha-2,3-sialytransferase); sialyltransferase ST3Gal-I; Siat4; SIAT4A; SIAT4-A; siat4-r1; SIATFL; ST3 beta-galactoside alpha-2,3-sialyltransferase 1; ST3Gal I; st3Gal I.1; ST3GAL1; St3gal-1; st3gal1A; st3gal1-r1; ST3GAL2; ST3GalA; ST3GalA.1; ST3GalI; ST3GAL-I sialyltransferase; ST3GalIA; ST3GalIA,1; ST3O |
| Símbolos de los genes | ST3GAL1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?