missing translation for 'onlineSavingsMsg'
Learn More

SSX1 Monoclonal Antibody (OTI2G5), TrueMAB™, OriGene

Product Code. 17577621
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Unit Size:
100 microlitros
30 microlitros
Product Code. Cantidad unitSize
17577621 30 μL 30 microlitros
17557621 100 μL 100 microlitros
2 options
This item is not returnable. View return policy

Product Code. 17577621

Brand: Origene Technologies TA502726S

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SSX1 Monoclonal antibody specifically detects SSX1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antígeno SSX1
Aplicaciones Flow Cytometry, Immunocytochemistry, Western Blot
Clasificación Monoclonal
Clon OTI2G5
Concentración 0.29 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica SSX1
N.º de referencia del gen Q16384
Alias de gen Cancer/testis antigen 5.1; cancer/testis antigen family 5, member 1; CT5.1; CT5.2; CT51; Protein SSX1; sarcoma, synovial, X-chromosome-related 1; SSRC; SSX family member 1; SSX1; SSX2; SSX2A; SSX2B; SSX3; Synovial sarcoma, X breakpoint 1; X breakpoint 2
Especie del huésped Mouse
Inmunógeno Full length human recombinant protein of human SSX1 produced in E.coli.
Método de purificación Affinity chromatography
Cantidad 30 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6756
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.