Learn More
SSSCA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12273-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The calculated MW of SSSCA1 is 21.5 kDa (native form), with modification the MW is about 27 kDa.
Might play a role in mitosis. Antigenic molecule. Could be a centromere-associated protein. May induce anti-centromere antibodies.Especificaciones
| SSSCA1 | |
| Polyclonal | |
| Unconjugated | |
| SSSCA1 | |
| Autoantigen p27, p27, SSSCA1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10534, 56390, 689397 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Flow Cytometry | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60232, P56873 | |
| Sssca1 | |
| SSSCA1 Fusion Protein Ag2916 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |