missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SSEA3 Monoclonal Antibody (MC-631), DyLight™ 488
Description
MA1-020-D488 has been successfully used in ICC/IF and flow cytometry applications on human samples. MA1-020-D488 can be used for immunofluoresence analysis of SSEA3 in human iPSCs.
SSEA-3 (stage-specific embryonic antigen 3) is a cell surface antigen, expressed along with SSEA-4, TRA-1-60 and TRA-1-81 in embryonic stem cells, embryonal carcinoma cells, and induced pluripotent stem cells (iPS). In human cells, these surface markers are down-regulated during the differentiation process, while SSEA1 is absent in undifferentiated human stem cells and upregulated on the cell surface after retinoic acid mediated differentiation. Conversely, in mouse stem cells, SSEA-1 expression decreases with differentiation while SSEA-3 and SSEA-4 expression increase.
Specifications
Specifications
| Antígeno | SSEA3 |
| Aplicaciones | ELISA, Flow Cytometry, Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | MC-631 |
| Concentración | 1 mg/mL |
| Conjugado | DyLight 488 |
| Formulación | PBS with proprietary stabilizer and 0.02% sodium azide |
| Alias de gen | SSEA-3; stage-specific embryonic antigen 3 |
| Especie del huésped | Rat |
| Inmunógeno | Fischer rat immunized with 4-8 cell stage mouse embryos. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?