missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-1 (CD15) Mouse, anti-Mouse, Alexa Fluor 488, Clone: MC-480, STEMCELL Technologies
Descripción
- The MC-480 antibody reacts with a terminal carbohydrate epitope, stage-specific embryonic antigen-1 (SSEA-1), which is expressed on a large-molecular-mass (> 200 kDa) glycoprotein on the surface of early mouse embryos, mouse embryonal carcinoma (EC), embryonic stem (ES) cells and mouse and human embryonic germ (EG) cells
- SSEA-1 is not expressed on undifferentiated human EC, ES or induced pluripotent stem (iPS) cells, or rhesus monkey ES cell lines
- It has been reported that SSEA-1 has roles in cell adhesion and migration, and regulation of cell differentiation
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ HLA Chimerism Whole Blood CD15 Positive Selection Kit (Catalog #17881; partial blocking may be observed), and for labeling human ES and iPS cells grown in TeSR™-E8™ (Catalog #05940), mTeSR™1 (Catalog #85850) and TeSR™2 (Catalog #05860)
Especificaciones
Especificaciones
| Antígeno | SSEA-1 (CD15) |
| Aplicaciones | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | MC-480 |
| Conjugado | Alexa Fluor 488 |
| Especie del huésped | Mouse |
| Cantidad | 25 Tests |
| Primario o secundario | Primary |
| Especies diana | Human, Rat, Mouse |
| Isotype | IgM κ |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?