missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SS18 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00€ - 468.00€
Especificaciones
| Antígeno | SS18 |
|---|---|
| Dilución | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
Descripción
SS18 Polyclonal antibody specifically detects SS18 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Especificaciones
| SS18 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 6760 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| MGC116875, Protein SYT, SSXTsynovial sarcoma, translocated to X chromosome, Synovial sarcoma translocated to X chromosome protein, synovial sarcoma translocation, chromosome 18, SYTprotein SSXT | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human SS18 (NP_001007560.1). MSVAFAAPRQRGKGEITPAAIQKMLDDNNHLIQCIMDSQNKGKTSECSQYQQMLHTNLVYLATIADSNQNMQSLLPAPPTQNMPMGPGGMNQSGPPPPPRSHNMPSDGMVGGGPPAPHMQNQMNGQMPGPNHMPMQGPGPNQLNMTNSSMNMPSSSHGSMGGYNHSVPSSQSMPVQNQMTMSQGQPMGNYGPRPNMSMQPNQGPMMHQQPPSQQYNMPQGGGQHYQGQQPPMGMMGQVNQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto