Learn More
SRY Mouse anti-Human, Unconjugated, Clone: 2E9A4, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67349-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The SRY antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SRY. This antibody recognizes human, rat antigen. The SRY antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
| SRY | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat |
| Western Blot, ELISA | |
| 2E9A4 | |
| Unconjugated | |
| 150 μL | |
| 6736 | |
| IgG1 |