missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR17018, MedChemExpress
Descripción
SR17018 is an mu-opioid-receptor (MOR) agonist, binding with GTPγS, with an EC50 of 97 nM.
Especificaciones
Especificaciones
| Nombre del producto químico o material | SR17018 |
| CAS | 2134602-45-0 |
| Color | Blanco |
| Notas de grado de pureza | Research |
| Fórmula molecular | C19H18Cl3N3O |
| Cantidad | 5 mg |
| Información de solubilidad | DMSO : 12.5 mg/mL (30.43 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C1NC2=CC(Cl)=C(Cl)C=C2N1C3CCN(CC4=CC=C(Cl)C=C4)CC3 |
| Peso molecular (g/mol) | 410.72 |
| Formula Weight (peso de la fórmula) | 410.72 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?