Learn More
SR-BI/SR-BII Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 9 publications
Marca:  Novus Biologicals NB400-134
 Ver más versiones de este producto
Código de producto. 18798453
  • ONLINE EXCLUSIVE PRICE
    492.00€
    465.15€ / 0.10 ml
    Ahorro 26.85€
    5% Descuento
Descripción
SR-BI/SR-BII Polyclonal specifically detects SR-BI/SR-BII in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen (Negative), Knockout Validated, Block/Neutralize.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
82 kDa
Unpurified
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Antisera
Certificados
Promociones
Promociones
For Research Use Only