missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR-BI Mouse anti-Human, Clone: 947007, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB8114-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SR-BI Monoclonal specifically detects SR-BI in Human samples. It is validated for Western Blot, Immunohistochemistry.Especificaciones
| SR-BI | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| SCARB1 | |
| Human embryonic kidney cell line HEK293-derived human SR-BI | |
| 25 μg | |
| Primary | |
| Detects human SR-BI in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 947007 | |
| Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL | |
| CD36 and LIMPII analogous 1, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1, CD36L1, CD36L1scavenger receptor class B type 1, CLA1, CLA1 CD36 antigen-like 1, CLA-1 HDLQTL6, Collagen type I receptor, thrombospondin receptor-like 1, HDLQTL6, MGC138242, SCARB1, scavenger receptor class B type III, scavenger receptor class B, member 1, SR-B1, SRB1 scavenger receptor class B member 1, SRBI | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 949 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?