missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SR-AI/MSR Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-00093CL1
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SR-AI/MSR Polyclonal antibody specifically detects SR-AI/MSR in Human,Mouse samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Especificaciones
| SR-AI/MSR | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| CD204, CD204 antigen, Macrophage acetylated LDL receptor I and II, macrophage scavenger receptor 1, macrophage scavenger receptor type III, phSR1, phSR2, SCARA1macrophage scavenger receptor types I and II, Scavenger receptor class A member 1, scavenger receptor class A, member 1, SR-A | |
| Synthetic peptide made to a cyclized peptide from human Macrophage Scavenger Receptor I (within residues 400-450). [Swiss-Prot# P21757] | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Immunology, Innate Immunity, Lipid and Metabolism, Neuroscience, Prostate Cancer | |
| 4481 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido