missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPEN Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA565605
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: GEAVENQEVQS KKPIPSKPQL KQLQVLDDQG PEREDVRKNY CSLRDETPER KSGQEKSHSV NTEEKIGIDI DHTQSYRKQM EQSRRKQQME MEIAKSEKFG SPKKDVDEYE RRSLVHEVGK PPQDVTDDSP PSK Highest antigen sequence identity to the following orthologs - mouse 78%, rat 82%.
May serve as a nuclear matrix platform that organizes and integrates transcriptional responses. In osteoblasts, supports transcription activation: synergizes with RUNX2 to enhance FGFR2-mediated activation of the osteocalcin FGF-responsive element (OCFRE). Has also been shown to be an essential corepressor protein, which probably regulates different key pathways such as the Notch pathway. Negative regulator of the Notch pathway via its interaction with RBPSUH, which prevents the association between NOTCH1 and RBPSUH, and therefore suppresses the transactivation activity of Notch signaling. Blocks the differentiation of precursor B-cells into marginal zone B-cells. Probably represses transcription via the recruitment of large complexes containing histone deacetylase proteins. May bind both to DNA and RNA.
Especificaciones
| SPEN | |
| Polyclonal | |
| Unconjugated | |
| SPEN | |
| HIAA0929; KIAA0929; MINT; Msx2 interacting nuclear target (MINT) homolog; msx2-interacting protein; nuclear receptor transcription cofactor; RBM15C; SHARP; SMART/HDAC1-associated repressor protein; SPEN; spen family transcriptional repressor; SPEN homolog; spen homolog, transcriptional regulator | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23013 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96T58 | |
| SPEN | |
| Recombinant Human SPEN. Recombinant protein control fragment (Product #RP-106268). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido