missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPATA25 Antibody (Z37P3H11*B2), CoraFluor™ 1, Novus Biologicals™
Descripción
SPATA25 Monoclonal antibody specifically detects SPATA25 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antígeno | SPATA25 |
| Aplicaciones | ELISA, Immunohistochemistry, Western Blot |
| Clasificación | Monoclonal |
| Clon | Z37P3H11*B2 |
| Conjugado | CoraFluor 1 |
| Formulación | PBS |
| Alias de gen | C20orf165, Chromosome 20 Open Reading Frame 165, DJ337O18.8, Spermatogenesis Associated 25, Testis-Specific Gene 23 Protein, TSG23 |
| Especie del huésped | Mouse |
| Inmunógeno | Peptide Sequence - LVRSKRGQP |
| Método de purificación | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?