Descripción
This antibody is specific to SPAG5.
SPAG5 (sperm associated antigen 5), also known as MAP126, hMAP126, DEEPEST or Astrin, is a 1,193 amino acid protein that is expressed at high levels in testis and at lower levels in liver, placenta, pancreas, colon and thymus. Localized specifically to spindle poles during prophase and throughout the spindle during metaphase and anaphase, SPAG5 is essential for proper spindle formation and is thought to be involved in the dynamic and functional regulation of mitotic spindles. Additionally, SPAG5 plays a role in sister chromatid cohesion and subsequent chromatid separation and, via its association with chromatids, is an important regulator of chromosome integrity. SPAG5 has a long stalk and a globular head domain through which if forms a homodimer that is characterized by aster-like structures. Upon DNA damage, SPAG5 may be phosphorylated by ATR or ATM.
Especificaciones
Especificaciones
| Antígeno | SPAG5 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation |
| Clasificación | Polyclonal |
| Concentración | 0.36 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | SPAG5 |
| N.º de referencia del gen | Q96R06 |
| Alias de gen | Astrin, DEEPEST, hMAP126, MAP126, SPAG5, sperm associated antigen 5 |
| Símbolos de los genes | SPAG5 |
| Mostrar más |