missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA541444
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Peptide sequence: GGTALAIVTS GELDSSVTEV LGSPRIVTVA AISQDSNPAT PNVSTNMEEF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
This gene belongs to a family of Sp1 related genes that encode transcription factors that regulate transcription by binding to consensus GC- and GT-box regulatory elements in target genes. This protein contains a zinc finger DNA-binding domain and several transactivation domains, and has been reported to function as a bifunctional transcription factor that either stimulates or represses the transcription of numerous genes. Transcript variants encoding different isoforms have been described for this gene, and one has been reported to initiate translation from a non-AUG (AUA) start codon. Additional isoforms, resulting from the use of alternate downstream translation initiation sites, have also been noted. A related pseudogene has been identified on chromosome 13.
Especificaciones
| SP4 | |
| Polyclonal | |
| Unconjugated | |
| Sp4 | |
| 5730497N03Rik; HF1B; HF-1b; HF1-b; HF-1b zinc finger protein; LOW QUALITY PROTEIN: transcription factor Sp4; MGC130008; MGC130009; SP4; Sp4 transcription factor; SPR-1; trans-acting transcription factor 4; transcription factor Sp4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6671 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q02446 | |
| Sp4 | |
| Synthetic peptide directed towards the middle region of human SP4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido