missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SP100 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA550076
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The antibody detects endogenous levels of total SP100 protein.
This gene encodes a subnuclear organelle and major component of the PML (promyelocytic leukemia)-SP100 nuclear bodies. PML and SP100 are covalently modified by the SUMO-1 modifier, which is considered crucial to nuclear body interactions. The encoded protein binds heterochromatin proteins and is thought to play a role in tumorigenesis, immunity, and gene regulation. Alternatively spliced variants have been identified for this gene; one of which encodes a high-mobility group protein.
Especificaciones
| SP100 | |
| Polyclonal | |
| Unconjugated | |
| SP100 | |
| A430075G10Rik; DKFZp686E07254; FLJ00340; FLJ34579; lysp100b; nuclear antigen Sp100; nuclear autoantigen Sp-100; Nuclear dot-associated Sp100 protein; SP 100; SP100; SP100 nuclear antigen; SP100-HMG nuclear autoantigen; speckled 100 kDa | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6672 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P23497 | |
| SP100 | |
| Synthesized peptide derived from Internal of human SP100. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido