Learn More
SOX10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Código de producto. 16816803 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
16816803 150 μL 150 microlitros
16826803 20 μL 20 microlitros
2 options
Código de producto. 16816803
Marca: Proteintech 104221AP150UL
Rabbit Polyclonal Antibody
SOX10 is a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. SOX10 plays a central role in developing and maturing of glia. Specifically, SOX10 activates expression of myelin genes (such as DUSPP15 and MYRF) during oligodendrocyte (OL) maturation; thus, playing a central role in oligodendrocyte maturation and CNS myelination. Once induced MYRF cooperates with SOX10 to implement the myelination program. SOX10 is also a transcriptional activator of MITF acting synergistically with PAX3. Mutations in this gene are associated with Waardenburg-Shah and Waardenburg-Hirschsprung disease.
Especificaciones
Antígeno SOX10
Aplicaciones Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Clasificación Polyclonal
Concentración 0.4 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
génica SOX10
N.º de referencia del gen O55170, P56693, Q04888
Alias de gen DOM, SOX10, Transcription factor SOX 10, WS2E, WS4
Símbolos de los genes SOX10
Especie del huésped Rabbit
Inmunógeno SOX10 Fusion Protein Ag0673
Método de purificación Antigen Affinity Chromatography
Cantidad 150 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 20665, 29361, 6663
Especies diana Human, Mouse, Rat
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG