missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOX10 Polyclonal Antibody

Descripción
Peptide sequence: PGGEAEQGGT AAIQAHYKSA HLDHRHPGEG SPMSDGNPEH PSGQSHGPPT Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 86%.
SOX10 is a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. This protein may act as a transcriptional activator after forming a protein complex with other proteins. It acts as a nucleocytoplasmic shuttle protein and is important for neural crest and peripheral nervous system development. Mutations in this gene are associated with Waardenburg-Shah and Waardenburg-Hirschsprung disease.
Especificaciones
Especificaciones
| Antígeno | SOX10 |
| Aplicaciones | ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Surface Plasmon Resonance |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 2% sucrose and 0.09% sodium azide |
| génica | SOX10 |
| N.º de referencia del gen | P56693 |
| Alias de gen | cls; cls/sox10; colorless; colourless; colourless/sox10; DOM; dominant megacolon; dominant megacolon, mouse, human homolog of; golas; gos; HMG-box transcription factor Sox10; MGC15649; PCWH; Protein SOX-21; SOX10; Sox-10; sox10 protein; sox10b; Sox21; SOX9; SRY; SRY (sex determining region Y)-box 10; SRY (sex determining region Y)-box 9 (campomelic dysplasia, autosomal sex-reversal); SRY box 10; SRY-box 10; SRY-box containing gene 10; SRY-box containing protein 10; SRY-box transcription factor 10; SRY-related HMG-box gene 10; transcription factor SOX-10; Transcription factor SOX-M; WS2E; WS4; WS4C; zgc:100757 |
| Símbolos de los genes | SOX10 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?