missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SORBS1 Polyclonal Antibody

Goat Polyclonal Antibody
Marca: Invitrogen™ PA518055
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is predicted to react with mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000. Preliminary analysis showed no signal in Western Blot.
Plays a role in tyrosine phosphorylation of CBL by linking CBL to the insulin receptor. Required for insulin-stimulated glucose transport. Involved in formation of actin stress fibers and focal adhesions.
Especificaciones
| SORBS1 | |
| Polyclonal | |
| Unconjugated | |
| SORBS1 | |
| 2310065E01Rik; 9530001P15Rik; CAP; c-Cbl associated protein; c-Cbl-associated protein; DKFZp451C066; DKFZP586P1422; Fas-ligand associated factor 2; FLAF2; FLJ12406; I79_015895; KIAA0894; KIAA1296; mKIAA1296; OTTHUMP00000020142; OTTHUMP00000020143; OTTHUMP00000020144; OTTHUMP00000020145; Ponsin; R85FL; SH3 domain protein 5; Sh3d5; SH3P12; SORB1; sorbin and SH3 domain containing 1; sorbin and SH3 domain-containing protein 1; Sorbs1; sorbs1 {ECO:0000250; UniProtKB:Q62417} | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 10580 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9BX66 | |
| SORBS1 | |
| Synthetic peptide sequence (GTFPGNYVKPLYL) corresponding to the C-terminus amino acids of SORBS1 (aa 1280-1292). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido