Learn More
Invitrogen™ Sonepcizumab Recombinant Monoclonal Antibody
Código de producto. 30284328
Change view
Click to view available options
Cantidad:
1 mg
100 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
30284328 1 mg 1 miligramo
30284017 100 μg 100 microgramos
2 options
Código de producto. 30284328
Marca: Invitrogen™ MA559796
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Sonepcizumab, also referred to as LT1009, is a humanized monoclonal antibody that targets sphingosine 1-phosphate (S1P), a signaling lipid active in promoting angiogenesis and tumor growth. Sphingosine-1-phosphate (S1P) is a critical lysolipid involved in multiple biological functions, including heart development, immune response, and hearing. It is a metabolite from cell membrane sphingolipids and is primarily regulated through its binding to specific G protein-coupled receptors named S1P receptors (S1PRs). These receptors are ubiquitously expressed in the human body, influencing various physiological and pathological processes. The signaling pathways initiated by S1P-S1PR interactions are crucial for immune cell trafficking, activation, and adaptive immune responses. Studies also highlight the role of S1P in regulating vascular stability, endothelial cell function, and as a mediator in inflammatory responses. The SPNS2 gene encodes a transporter protein essential for the active secretion of S1P, further emphasizing its relevance in maintaining physiological homeostasis.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Sonepcizumab Humanized
Aplicaciones ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Clasificación Recombinant Monoclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación 25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Especie del huésped Human
Método de purificación Protein A
Cantidad 1 mg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG1