Descripción
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This endosomal protein regulates the cell-surface expression of epidermal growth factor receptor. This protein also has a role in sorting protease-activated receptor-1 from early endosomes to lysosomes. This protein may form oligomeric complexes with family members. This gene results in three transcript variants encoding distinct isoforms.
Especificaciones
Especificaciones
| Antígeno | SNX1 |
| Aplicaciones | Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Concentración | 0.2 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | SNX1 |
| N.º de referencia del gen | Q13596, Q99N27, Q9WV80 |
| Alias de gen | HsT17379, SNX1, SNX1A, sorting nexin 1, Vps5 |
| Símbolos de los genes | Snx1 |
| Mostrar más |