missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMYD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA587309
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Purity is > 95% by SDS-PAGE.
SMYD3 (histone-lysine N-methyltransferase) methylates Lys-4 of histone H3 and Lys-5 of histone H4. It induces di- and tri-methylation. SMYD3 plays an important role in transcriptional activation as a member of an RNA polymerase complex.
Especificaciones
| SMYD3 | |
| Polyclonal | |
| Unconjugated | |
| SMYD3 | |
| 2410008A19Rik; bA74P14.1; bA74P14.1 (novel protein); histone-lysine N-methyltransferase SMYD3; KMT3E; SET and MYND domain containing 3; SET and MYND domain-containing protein 3; Smyd3; Zinc finger MYND domain-containing protein 1; zinc finger protein, subfamily 3A (MYND domain containing), 1; zinc finger, MYND domain containing 1; ZMYND1; ZNFN3A1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64754, 69726 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9CWR2, Q9H7B4 | |
| SMYD3 | |
| Recombinant full length Human SMYD3. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido