Learn More
Smooth Muscle Actin Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Código de producto. 17244676 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
17244676 100 μL 100 microlitros
1 options
Código de producto. 17244676
Marca: Proteintech CL48814395100UL
Rabbit Polyclonal Antibody
ACTA2 (also known as α-smooth muscle actin or α-SMA) belongs to the actin family. Actins are highly conserved proteins that are involved in various types of cell motility and are ubiquitously expressed in all eukaryotic cells. ACTA2 is primarily expressed in vascular smooth muscle and anti-ACTA2 is commonly used to marker smooth muscle cells. This antibody may cross-react with other isoforms of actins.
Especificaciones
Antígeno Smooth Muscle Actin
Aplicaciones Immunofluorescence
Clasificación Polyclonal
Concentración 1000 μg/mL
Conjugado CoraLite 488
Especie del huésped Rabbit
Cantidad 100 μL
Primario o secundario Primary
ID de gen (Entrez) 59
Especies diana Human, Mouse, Rat, Pig
Formulario Liquid
Isotype IgG