Learn More
SMG5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12694-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The SMG5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SMG5. This antibody recognizes human antigen. The SMG5 antibody has been validated for the following applications: ELISA analysis.
Especificaciones
| SMG5 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 23381 | |
| IgG |
| Western Blot, ELISA | |
| 133 μg/mL | |
| Rabbit | |
| Primary | |
| Human |