missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMC2 Polyclonal Antibody
Descripción
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
CAP-E and CAP-C (Chromosome Associated Protein-E & C) are also SMC (Structural Maintenance of Chromosome) family members that form a heterodimeric complex required for mitotic chromosome condensation to achieve proper segregation of genetic information during subsequent cell division. hCAP-C/hCAP-E is a component of a multiprotein complex called condensin that interacts with phosphorylation of Histone H3 resulting in the mitotic chromosome condensation. The distribution patterns of the two heterodimeric complexes in interphase nucleus indicate independent behavior of the two complexes during cell cycle. These results suggest that the two distinct complexes are involved in different aspects of mitotic chromosome organization in humar cells.
Especificaciones
Especificaciones
| Antígeno | SMC2 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| génica | SMC2 |
| N.º de referencia del gen | O95347 |
| Alias de gen | 5730502P04Rik; AI255214; AW545314; CAP E; CAPE; CAP-E; Chromosome-associated protein E; FGF-inducible protein 16; fibroblast growth factor inducible 16; Fin16; FLJ10093; hCAP-E; PRO0324; SMC protein 2; Smc2; SMC-2; SMC2 (structural maintenance of chromosomes 2, yeast)-like 1; SMC2 protein; SMC2 structural maintenance of chromosomes 2-like 1; Smc2l1; structural maintenance of chromosomes (SMC) family member, chromosome-associated protein E; structural maintenance of chromosomes 2; structural maintenance of chromosomes protein 2; XCAP-E homolog |
| Símbolos de los genes | SMC2 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?