missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
The protein encoded by this gene is a member of the SWI/SNF family of proteins and is similar to the brahma protein of Drosophila. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI, which is required for transcriptional activation of genes normally repressed by chromatin. In addition, this protein can bind BRCA1, as well as regulate the expression of the tumorigenic protein CD44. Multiple transcript variants encoding different isoforms have been found for this gene.
Especificaciones
Especificaciones
| Antígeno | SMARCA4 |
| Aplicaciones | Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation |
| Clasificación | Monoclonal |
| Clon | 2E6B6 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | SMARCA4 |
| N.º de referencia del gen | P51532, Q8K1P7 |
| Alias de gen | ATP dependent helicase SMARCA4, BAF190, BAF190A, BRG1, BRG1 associated factor 190A, hSNF2b, Protein brahma homolog 1, Protein BRG 1, SMARCA4, SNF2, SNF2 BETA, SNF2B, SNF2L4, SNF2LB, SWI2, Transcription activator BRG1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?