Learn More
SMAD7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25840-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The SMAD7 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human SMAD7. This antibody recognizes human, mouse, rat antigen. The SMAD7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Especificaciones
| SMAD7 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 4092 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
| 540 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |