missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMAD5 Recombinant Rabbit Monoclonal Antibody (1X5G9)

Descripción
Immunogen sequence: STYPNSPASS GPGSPFQLPA DTPPPAYMPP DDQMGQDNSQ PMDTSNNMIP QIMPSISSRD VQPVAYEEPK HWCSIVYYEL NNRVGEAFHA SSTSVLVDGF T.
SMADs are members of the MAD-related family of molecules. MAD-related proteins are a family of intracellular proteins that are essential components in the signaling pathways of the serine/threonine kinase receptors of the transforming growth factor beta superfamily. SMADs can be divided into receptor-regulated SMADs (R-SMADs: SMAD1, 2, 5, 8 and 9), common-mediator SMAD (co-SMAD: SMAD4), and inhibitory SMADs (I-SMADs: SMAD6 and 7). SMAD1, 5, 8 and 9 have high degrees of homology and antibodies are available that recognize sequences common to all of them. SMAD8 and SMAD9 are typically used as alternate names for one another in the literature.
Especificaciones
Especificaciones
| Antígeno | SMAD5 |
| Aplicaciones | ELISA, Immunoprecipitation, Western Blot |
| Clasificación | Recombinant Monoclonal |
| Clon | 1X5G9 |
| Concentración | 0.75 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| génica | SMAD5 |
| N.º de referencia del gen | P97454, Q99717 |
| Alias de gen | 1110051M15Rik; AI451355; DKFZp781C1895; DKFZp781O1323; dwarfin-C; DWFC; Dwf-C; hSmad5; JV5-1; MAD homolog 5; MAD, mothers against decapentaplegic homolog 5; Madh5; Mothers against decapentaplegic homolog 5; mothers against decapentaplegic, drosophila, homolog of, 5; mothers against DPP homolog 5; mSmad5; MusMLP; OTTHUMP00000223331; SMA- and MAD-related protein 5; SMAD; SMAD 5; SMAD family member 5; SMAD, mothers against DPP homolog 5; SMAD5 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?