missing translation for 'onlineSavingsMsg'
Learn More

SMAD2 Monoclonal Antibody (5G7), Invitrogen™

Código de producto. 12807902
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
12807902 100 μL 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 12807902 Proveedor Invitrogen N.º de proveedor MA515877

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

MA5-15877 targets SMAD2 in FACS, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15877 immunogen is purified recombit fragment of human SMAD2 expressed in E. Coli. . MA5-15877 detects SMAD2 which has a predicted molecular weight of approximately 52kDa.

SMAD2 (MADH2, MAD2) regulates multiple cellular processes, such as cell proliferation, apoptosis, and differentiation. SMAD2 interacts with the TGF-beta receptors through its interaction with the SMAD anchor for receptor activation into the nucleus is a central event in TGF beta signaling. Phosphorylation of threonine 8 in the calmodulin-binding region of the MH1 domain by extracellular signal regulated kinase 1 (ERK1) enhances SMAD2 transcriptional activity, which is negatively regulated by calmodulin. In response to the TGF-beta signal, SMAD2 is phosphorylated by the TGF-beta receptors. The phosphorylation induces the dissociation of this protein with SARA and the association with the family member SMAD4. The association with SMAD4 is important for the translocation of this protein into the nucleus, where it binds to target promoters and forms a transcription repressor complex with other cofactors. SMAD2 can also be phosphorylated by activin type 1 receptor kinase, and mediates the signal from the activin. Alternatively spliced transcript variants encoding the SMAD2 protein have been observed.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno SMAD2
Aplicaciones Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Clasificación Monoclonal
Clon 5G7
Conjugado Unconjugated
génica SMAD2
N.º de referencia del gen Q15796, Q62432
Alias de gen JV18, JV18-1, MADH2, MADR2, hMAD-2, hSMAD2, MAD homolog 2; SMAD, mothers against DPP homolog 2; Sma- and Mad-related protein 2; mother against DPP homolog 2; mothers against decapentaplegic homolog 2
Símbolos de los genes SMAD2
Especie del huésped Mouse
Inmunógeno Purified recombit fragment of human SMAD2 expressed in E. Coli.
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 17126, 4087
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.