missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Slo2.2 Monoclonal Antibody (N3/26), PE
GREENER_CHOICE

Código de producto. 17827472
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17827472 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17827472 Proveedor Invitrogen™ N.º de proveedor MA545437

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

1 μg/mL of MA5-45437 was sufficient for detection of Slo2.2 in 10 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 140kDa. Weak human detection.approximately Does not cross-react with KCNT2/Slo2.1/Slick.

Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a sodium-activated potassium channel subunit which is thought to function in ion conductance and developmental signaling pathways. Mutations in this gene cause the early-onset epileptic disorders, malignant migrating partial seizures of infancy and autosomal dominant nocturnal frontal lobe epilepsy. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Slo2.2
Aplicaciones Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Microarray
Clasificación Monoclonal
Clon N3/26
Concentración 1 mg/mL
Conjugado PE
Formulación 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
génica Kcnt1
N.º de referencia del gen Q5JUK3, Q6ZPR4, Q9Z258
Alias de gen bA100C15.2; C030030G16Rik; EIEE14; ENFL5; KCa4.1; KCa4.1KIAA1422FLJ41282; Kcnt1; KIAA1422; Potassium channel subfamily T member 1; potassium channel subunit (Slack); potassium channel, sodium activated subfamily T, member 1; potassium channel, sodium-activated subfamily T, member 1; potassium channel, subfamily T, member 1; potassium sodium-activated channel subfamily T member 1; rSlo2; Sequence like a calcium-activated K+ channel; sequence like a calcium-activated potassium channel subunit; Slack; slo2; Slo2.2
Símbolos de los genes Kcnt1
Especie del huésped Mouse
Inmunógeno Fusion protein amino acids 1168-1237 of rat Slo2.2 (Slack).
Método de purificación Protein G
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 227632, 57582, 60444
Especies diana Human, Mouse, Rat
Contenido y almacenamiento 4°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG1
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.