missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLITRK6 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-76877PCP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
SLITRK6 Polyclonal antibody specifically detects SLITRK6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Especificaciones
| SLITRK6 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| FLJ22774, MGC119595, MGC119596, MGC119597,4832410J21Rik, SLIT and NTRK-like family, member 6, SLIT and NTRK-like protein 6, slit and trk like gene 6 | |
| Antibody was raised against a 13 amino acid synthetic peptide from near the center of human Slitrk6. The immunogen is located within amino acids 250 - 300 of Slitrk6. Amino Acid Squence: TTPVYEEHEDPSG | |
| 0.1 mL | |
| Neuroscience | |
| 84189 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido