Learn More
Ibidi μ-Slide 8 Well High Glass Bottom
Descripción
- A chambered coverslip with 8 individual wells and high walls
- suitable for use in TIRF and super-resolution microscopy
- bottom material: #1.5H (170 μm ± 5 μm) D 263 M Schott glass coverslip
- sterilized
Especificaciones
Especificaciones
| Tipo | Chambered Coverglass |
| Pocillos | 8 |