missing translation for 'onlineSavingsMsg'
Learn More

SLFN12 Antibody, CoraFluor™ 1, Novus Biologicals™

Product Code. 30146624
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
30146624 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 30146624

Brand: Novus Biologicals (Bio-Techne) NBP176252CL1

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLFN12 Polyclonal antibody specifically detects SLFN12 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno SLFN12
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Conjugado CoraFluor 1
Formulación PBS
Alias de gen schlafen family member 12
Especie del huésped Rabbit
Inmunógeno Antibody was raised against a 12 amino acid synthetic peptide from near the carboxy terminus human SLFN12. The immunogen is located within the last 50 amino acids of SLFN12. Amino Acid Squence: FQKNDKKMFKS
Método de purificación Peptide affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
ID de gen (Entrez) 55106
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark. Do not freeze.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.