missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC4A1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 28131-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The Band 3 protein is the most abundant integral protein of human erythrocyte membranes and functions as the major anion transporter polypeptide of erythrocytes. Band 3 is a 90-100 kDa protein, its microheterogeneity is ascribed mainly to the structural heterogeneity of the carbohydrate moiety. The C-terminal portion of the molecule spans the membrane several times, and is responsible for anion transport. The N-terminal portion, comprising almost half of the polypeptide chain of Band 3, is located inside the red cell and interacts with cytoskeletal and cytoplasmic proteins such as ankyrin.Especificaciones
| SLC4A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC4A1 | |
| AE 1, AE1, Anion exchange protein 1, Anion exchanger 1, Band 3 anion transport protein, BND3, CD233, DI, EMPB3, EPB3, FR, RTA1A, SLC4A1, SW, WD, WD1, WR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20533, 6521 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P02730, P04919 | |
| SLC4A1 | |
| SLC4A1 Fusion Protein Ag27878 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido