missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC47A2 Polyclonal Antibody

Código de producto. 13211819
Change view
Click to view available options
Cantidad:
400 μL
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
13211819 400 μL 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 13211819

Marca: Invitrogen™ PA524659

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

SLC47A2 Polyclonal Antibody for Western Blot

S47A2 belongs to a family of transporters involved in excretion of toxic electrolytes, both endogenous and exogenous, through urine and bile. This transporter family shares homology with the bacterial MATE (multidrug and toxin extrusion) protein family responsible for drug resistance. This gene is one of two members of the MATE transporter family located near each other on chromosome 17.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno SLC47A2
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.5 mg/mL
Conjugado Unconjugated
Formulación PBS with 0.09% sodium azide; pH 7.4
génica Slc47a2
N.º de referencia del gen Q86VL8
Alias de gen 4933429E10Rik; FLJ31196; H(+)/organic cation antiporter kidney-specific; hMATE-2; kidney-specific H(+)/organic cation antiporter; Mate2; MATE-2; MATE2-B; MATE2K; MATE2-K; mMATE-2; multidrug and toxin extrusion protein 2; SLC47A2; solute carrier family 47 (multidrug and toxin extrusion), member 2; solute carrier family 47 member 2; solute carrier family 47, member 2
Símbolos de los genes Slc47a2
Especie del huésped Rabbit
Inmunógeno KLH conjugated synthetic peptide between 125-153 amino acids from the N-terminal region of human S47A2.
Método de purificación Antigen Affinity Chromatography, Protein A
Cantidad 400 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 146802, 497921
Especies diana Human, Rat
Contenido y almacenamiento Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.