Learn More
Invitrogen™ SLC44A4 Biosimilar Recombinant Monoclonal Antibody
GREENER_CHOICE
Código de producto. 30283845
Change view
Click to view available options
Cantidad:
1 mg
100 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
30283845 100 μg 100 microgramos
30283555 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 30283845 Proveedor Invitrogen™ N.º de proveedor MA559001
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term ASG-5ME.
SLC44A4 (solute carrier family 44 member A4) is a gene encoding a protein that primarily functions as the thiamine pyrophosphate transporter (TPPT) in humans. This protein is responsible for the absorption of thiamine pyrophosphate (TPP), a major form of vitamin B1, generated by gut microbiota in the large intestine. The SLC44A4 gene is highly expressed in the colon and is localized to the apical membrane of colonic cells, enabling efficient uptake of TPP. This transporter operates in a sodium-independent manner and is crucial for maintaining thiamine homeostasis, which is vital for various cellular processes including carbohydrate metabolism and neural function. Disruption or mutations in SLC44A4 can impact the body's ability to absorb thiamine, leading to deficiencies that may result in metabolic and neurological disorders.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno SLC44A4 Biosimilar
Aplicaciones ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Clasificación Recombinant Monoclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Especie del huésped Human
Método de purificación Protein A
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
Especies diana Human
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Tipo de producto Antibody
Formulario Lyophilized
Isotype IgG2SA