missing translation for 'onlineSavingsMsg'
Learn More

SLC38A1 Rabbit Polyclonal Antibody, Proteintech

Código de producto. 30233588 Tienda Proteintech Productos
Change view
Click to view available options
Cantidad:
20 μL
150 μL
Tamaño de la unidad:
150 microlitros
20 microlitros
Código de producto. Cantidad unitSize
30233588 20 μL 20 microlitros
30235763 150 μL 150 microlitros
2 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 30233588

missing translation for 'mfr': Proteintech 286321AP20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The SLC38A1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC38A1. This antibody recognizes human, mouse antigen. The SLC38A1 antibody has been validated for the following applications: WB, IHC, ELISA analysis.

Specifications

Antígeno SLC38A1
Aplicaciones Western Blot, Immunohistochemistry, ELISA
Clasificación Polyclonal
Concentración 500 μg/mL
Conjugado Unconjugated
Símbolos de los genes ATA1; NAT2; SAT1; SNAT1
Especie del huésped Rabbit
Inmunógeno Recombinant Protein
Método de purificación Antigen affinity purification
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 81539
Especies diana Human, Mouse
Contenido y almacenamiento Store at -20°C. Stable for one year after shipment.
Peso molecular del anticuerpo 54 kDa, 60-70 kDa
Tipo de producto Polyclonal Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.