missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC35C2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5100800
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Antibody detects endogenous levels of total SLC35C2.
May play an important role in the cellular response to tissue hypoxia. May be either a GDP-fucose transporter that competes with SLC35C1 for GDP-fucose, or a factor that otherwise enhances the fucosylation of Notch and is required for optimal Notch signaling in mammalian cells.
Especificaciones
| SLC35C2 | |
| Polyclonal | |
| Unconjugated | |
| SLC35C2 | |
| BA394O2.1; C20orf5; C85957; CGI-15; D2Wsu58e; MGC137415 protein; ovarian cancer overexpressed 1; ovarian cancer overexpressed gene 1 protein; ovarian cancer-overexpressed gene 1 protein; Ovcov1; Slc35c2; slc35c2 protein; solute carrier family 35 (GDP-fucose transporter), member C2; solute carrier family 35 member C2; solute carrier family 35, member C2; wu:fb99e07; wu:fc49g11; wu:fi09e02; zgc:55285; zgc:77328 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 228875, 51006 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8VCX2, Q9NQQ7 | |
| SLC35C2 | |
| A synthesized peptide derived from human SLC35C2(Accession Q9NQQ7), corresponding to amino acid residues D324-Q365. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido